missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Product Code. 11330068
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330068 2 g 2g
11320068 10 g 10g
11310068 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11330068 Supplier Thermo Scientific Alfa Aesar Supplier No. 042844.04
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plays an important role in the production of dry cell batteries. | CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
IUPAC Name manganese(2+);dichloride
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190°C
Melting Point 650°C
Physical Form Powder
Quantity 2 g
Packaging Ampouled under argon
Sensitivity Hygroscopic
Specific Gravity 2.977
Density 2.977
Linear Formula MnCl2
UN Number UN3288
Solubility Information Soluble in water,pyridine and alcohol. Insoluble in ether.
Formula Weight 125.84
Percent Purity 99.99%
Chemical Name or Material Manganese(II) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.