missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Nitrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | 37°C |
| CAS | 20694-39-7 |
| Color | Red |
| Physical Form | Solid |
| Quantity | 500 g |
| Molecular Formula | H8MnN2O10 |
| MDL Number | 11112 |
| Synonym | manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate |
| InChI Key | ALIMWUQMDCBYFM-UHFFFAOYSA-N |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2] |
| Show More |
Safety and Handling
Product Identifier
- Manganese(II) nitrate tetrahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?