missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)

Product Code. 11498420
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498420 50 g 50g
11488420 10 g 10g
11478420 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498420 Supplier Thermo Scientific Alfa Aesar Supplier No. 044318.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol

Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15364-94-0
Molecular Formula Cl2H12MnO14
Molecular Weight (g/mol) 361.92
MDL Number MFCD00150255
InChI Key AIFGVOMWPFMOCN-UHFFFAOYSA-L
Synonym manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade
PubChem CID 203855
IUPAC Name manganese(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 165°C (decomposition)
Physical Form Crystalline
Quantity 50 g
Odor Odorless
Sensitivity Hygroscopic
UN Number UN1481
Solubility Information Soluble in water,benzene,ethyl acetate
Formula Weight 361.93 (253.84 Anhydrous)
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Manganese (II) perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.