missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) perchlorate hexahydrate, 99%

Product Code. 10155570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155570 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155570 Supplier Thermo Scientific Acros Supplier No. 316511000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 165°C
CAS 15364-94-0
Color Pink
Physical Form Crystalline Solid
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula Cl2H12MnO14
Linear Formula Mn(ClO4)2·6H2O
MDL Number MFCD00150255
Synonym manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade
Solubility Information Solubility in water: soluble in water
InChI Key AIFGVOMWPFMOCN-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
IUPAC Name manganese(2+);diperchlorate;hexahydrate
Molecular Weight (g/mol) 361.92
PubChem CID 203855
Formula Weight 361.93
Percent Purity 99%
Chemical Name or Material Manganese(II) perchlorate hexahydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(II) perchlorate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Manganese(II) perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.