missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, Analytical Reagent, Honeywell Fluka™

Product Code. 16625803 Shop All Honeywell Fluka Products
Change view
Click to view available options
:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16625803 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625803 Supplier Honeywell Fluka Supplier No. 3142525KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 700°C (anhydrous substance)
CAS 10034-96-5
Color Pink
CAS Max % 1
pH 4 to 6
Quantity 25 kg
Molecular Formula H2MnO5S
Linear Formula MnSO4 · H2O
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
Grade ACS
Density 2.950 g/cm3 (20°C)
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.