missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, Puriss., meets analytical spec. of Ph. Eur, BP, USP, FCC, 99 to 100.5% (calc. for dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15691280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
50 kg
Unit Size:
1kg
2.5kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15691280 50 kg 50kg
15671280 1 kg 1kg
15681280 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691280 Supplier Honeywell-Fluka Supplier No. 1324550KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100.0000%
Quantity 50 kg
Assay Percent Range 99 to 100.5%
Linear Formula MnSO4 · H2O
UN Number UN3077
Formula Weight 169.02
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Fiberboard Box
Density 2.950 g/cm3
Chemical Name or Material Manganese(II) sulfate monohydrate, Puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.