missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%, Solstice

Product Code. 15633400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
6 x 1 kg
Unit Size:
1kg
500g
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15633400 6 x 1 kg Pack of 6
15613400 500 g 500g
15603400 1 kg 1kg
15623400 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15633400 Supplier Honeywell-Fluka Supplier No. 314256X1KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Quantity 6 x 1 kg
Linear Formula MnSO4 · H2O
UN Number UN3077
Formula Weight 169.02
Percent Purity 99.0 to 101.0%
Grade ACS/puriss. p.a.
Chemical Name or Material Manganese(Ii) Sulfate Monohydrate, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.