missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulphate monohydrate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10665892 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10665892 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665892 Supplier JT Baker Supplier No. 0174.1000
  • ONLINE EXCLUSIVE PRICE
    152.00 €
    132.79 € / 1kg
    Save 19.21 €
    12% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Exceeds ACS Specifications

Specifications

Melting Point 700°C
CAS 10034-96-5
Quantity 1 kg
Assay Percent Range 98.0 to 101.0%
Molecular Formula H2MnO5S
MDL Number MFCD00149159
Synonym Manganous sulphate monohydrate , Manganese sulphate monohydrate
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
Loss on Ignition 10.0 - 12.0%
Packaging Plastic bottle for solids
Density 2.95 g/cm³ (25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.