missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) fluoride, 98% (metals basis)

Product Code. 11332168
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332168 50 g 50g
11322168 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332168 Supplier Thermo Scientific Alfa Aesar Supplier No. 013069.18
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-53-1

It is used as a fluorinating agent, useful for conversion of hydrocarbons into fluorocarbons. It is used in organic synthesis as Lewis acid to prepare a number of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-53-1
MDL Number MFCD00016223

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 50 g
Sensitivity Hygroscopic
Density 3.54 g/mL
UN Number UN3288
Merck Index 14,5741
Solubility Information Insoluble in organic solvents. Hydrolyzed by water.
Formula Weight 111.93
Percent Purity 98%
Assay (metals basis)
Chemical Name or Material Manganese(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.