missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99.99+%, (trace metal basis)

Product Code. 10259643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Packaging Förpackningsstorlek
10259643 25 g Glass bottle 25g
10626402 5 g Glass bottle 5g
13448438 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 10259643 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 193470250
 Restricted Item
In Stock
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Oxidation reagent
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Tekniske data

Melting Point 535.0°C
Color Black to Gray
Physical Form Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 99.99+%
Linear Formula MnO2
Fieser 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
Merck Index 15,5795
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Formula Weight 86.94
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Density 5.02g/mL
Chemical Name or Material Manganese(IV) oxide
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.