missing translation for 'onlineSavingsMsg'
Learn More

Manganese(Iv) Oxide, Technical, Powder, Fisher Chemical

Product Code. 10234440 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10234440 500 g Glass Jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234440 Supplier Fisher Chemical Supplier No. M/1660/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 535°C
CAS 1313-13-9
Color Gray
Physical Form Solid
Quantity 500 g
Molecular Formula MnO2
MDL Number MFCD00003463
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILES O=[Mn]=O
IUPAC Name dioxomanganese
Molecular Weight (g/mol) 86.94
PubChem CID 14801
Formula Weight 86.94g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(IV) oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.