missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mn | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 7439-96-5, 8042-47-5 |
CAS Min % | 0.5, 99.5 |
CAS Max % | 0.5, 99.5 |
Molecular Formula | Mn |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
SMILES | [Mn] |
IUPAC Name | manganese |
Molecular Weight (g/mol) | 54.94 |
PubChem CID | 23930 |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'