missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals

Product Code. 11411921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411921 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411921 Supplier MP Biomedicals Supplier No. 0219470290
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Crystalline grade of manganese sulfate monohydrate suitable for general laboratory research.

Specifications

CAS 10034-96-5
Physical Form Crystalline
Quantity 500 g
Molecular Formula H2MnO5S
MDL Number MFCD00149159
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
PubChem CID 177577
ChEBI CHEBI:86364
Formula Weight 169
Grade Cell Culture
Chemical Name or Material Manganese Sulfate Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.