missing translation for 'onlineSavingsMsg'
Learn More

Manninotriose, MedChemExpress

Product Code. 16471598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16471598

Brand: Medchem Express HYN0913

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Manninotriose is a novel and important player in the RFO(Raffinose family oligosaccharides) metabolism of red dead deadnettle; potential to improve the side effects of MTX for ALL treatment.

Manninotriose is a novel and important player in the RFO(Raffinose family oligosaccharides) metabolism of red dead deadnettle

Specifications

Chemical Name or Material Manninotriose
CAS 13382-86-0
Color White
Purity Grade Notes Research
Molecular Formula C18H32O16
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (198.24 mM; Need ultrasonic) ∣H2O : 83.3 mg/mL (165.13 mM; Need ultrasonic and warming)
SMILES O[C@@H]([C@@H](O)[C@H](O)[C@@H](O)C=O)CO[C@H]([C@@H]([C@@H](O)[C@H]1O)O)O[C@@H]1CO[C@H]([C@@H]([C@@H](O)[C@H]2O)O)O[C@@H]2CO
Molecular Weight (g/mol) 504.44
Formula Weight 504.44
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.