missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannitol, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
500g, Plastic powder jar
Specifications
Specifications
| CAS | 69-65-8 |
| Melting Point | 165°C |
| Color | White |
| Packaging | Plastic powder jar |
| Molecular Formula | C6H14O6 |
| MDL Number | 64287 |
| Quantity | 500 g |
| Formula Weight | 182.17g/mol |
| Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction