missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15558024 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15558024 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558024 Supplier Avantor J.T.Baker Supplier No. 255401
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 69-65-8
Packaging Poly Bottle
Molecular Formula C6H14O6
MDL Number MFCD00064287
Quantity 500 g
Specific Gravity 1L = 1.52kg
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES OCC(O)C(O)C(O)C(O)CO
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
PubChem CID 6251
ChEBI CHEBI:16899
Formula Weight 182.17
Grade Not Available
Physical Form Powder
Chemical Name or Material Mannitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.