missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Mannitol Selenite Broth Base
Click to view available options
:
Mannitol Selenite Broth Base 500g
Description
Mannitol Selenite Broth deydrated culture medium is a modification of Selenite F Broth for the selective enrichment of salmonellae
Formula: Bacteriological peptone 5.0 gm/litreMannitol 4.0 gm/litreSodium phosphate 10.0 gm/litrepH 7.1 ± 0.2Requires LP0121A Sodium Biselenite 100g
Specifications
Specifications
| Content And Storage | Room Temperature |
| Country of Origin | UK |
| Media Type | Dehydrated Culture Medium |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Enrichment of Salmonellae |
| Packaging Type | Tub |
| Quantity | 500 g |
| Species | Salmonella species |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction