missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals

Product Code. 11801613 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
For Flip Tear Up
For Flip Top/for Flip Off
Diameter (Metric) Cap:
13 mm
20 mm
28 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Cap unitSize
11801613 For Flip Top/for Flip Off 20 mm Each
11844970 For Flip Tear Up 13 mm Each
11829440 For Flip Top/for Flip Off 28 mm Each
11875050 For Flip Tear Up 20 mm Each
11777597 For Flip Top/for Flip Off 13 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11801613
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 31-07-2026
Add to basket
Add to basket

Crimping tools provide a reproducible, secure vial closure.

  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY

Specifications

Description 20mm Crimper
Diameter (Metric) Cap 20 mm
Material Steel
For Use With (Equipment) For Flip Top/for Flip Off
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.