missing translation for 'onlineSavingsMsg'
Learn More

MAP1LC3A 293T Cell Overexpression Lysate (Denatured), Abnova

Product Code. 16195953
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16195953 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16195953 Supplier Abnova Supplier No. H00084557T01.100uL
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell transient overexpression lysate for MAP1LC3A

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B.

  • Transfected cell line: 293T
  • Plasmid: pCMV-MAP1LC3A full-length
  • Host: Human
  • Theoretical molecular weight: 13.42kDa
  • Storage buffer: 1X sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description MAP1LC3A 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC015810
Molecular Weight (g/mol) 13.31
pH Range 6.8
Plasmid pCMV-MAP1LC3A full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-MAP1LC3A antibody (H00084557-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.