missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline Hydrochloride, TRC

Product Code. 30471728
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471728 Supplier Toronto Research Chemicals Supplier No. TRCM1880005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maprotiline Hydrochloride
CAS 10347-81-6
Molecular Formula C20 H23 N . Cl H
Quantity 5 g
Synonym 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (1:1), 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (9CI), 9,10-Ethanoanthracene-9(10H)-propylamine, N-methyl-, hydrochloride (7CI,8CI), 9,10-Dihydro-9-(3-methylaminopropyl)-9,10-ethanoanthracene hydrochloride, 9,10-Ethanoanthracene-9-propylamine, 9,10-dihydro-N-methyl-, hydrochloride, 9-[γ-(Methylamino)propyl]-9,10-dihydro-9,10-ethanoanthracene hydrochloride, Ba 34276, CIBA 34,276Ba, CIBA 34276 hydrochloride, Deprilept, Ladiomil, Ludiomil, Maprotiline hydrochloride, Psymion
InChI Formula InChI=1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H
SMILES Cl.CNCCCC12CCC(c3ccccc13)c4ccccc24
IUPAC Name N-Methyl-9,10-ethanoanthracene-9(10H)-propanamine hydrochloride
Molecular Weight (g/mol) 313.86
Formula Weight 313.1597
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.