missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Margatoxin, 99+%

Product Code. p-7070070
Change view
Click to view available options
Quantity:
0.1 mg
5 x 10 μg
5 x 5 μg
Unit Size:
0.1mg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423759 5 x 5 μg Each
15413759 5 x 10 μg Each
15433759 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423759 Supplier Thermo Scientific™ Supplier No. J61103.!X
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Description White
Quantity 5 x 5 μg
Assay 99+%
CAS 145808-47-5
Molecular Formula C178H286N52O50S7
Formula Weight 4178.95
Synonym rMargatoxin; MgTx
Product Type Margatoxin
Content And Storage Ambient temperatures
For Use With (Application) Peptide that blocks voltage-dependent Kv1.3 potassium channels
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.