Learn More
Emma Safety Footwear Martino (Frontier 162) Safety Shoes
Product Code. 15938271
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15938271 43 1 pair
15908271 39 1 pair
15918271 40 1 pair
15928271 41 1 pair
15805781 42 1 pair
15948271 44 1 pair
15958271 45 1 pair
15968271 46 1 pair
15978271 47 1 pair
15988271 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15938271 Supplier Emma Safety Footwear Supplier No. MM16209043
  • ONLINE EXCLUSIVE PRICE
    247.80 €
    155.40 € / 1 pairs
    Save 92.40 €
    37% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The executive safety shoe is made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
  • S3-Safety shoe with sole plate for wet work
  • Thermoplastic Polyurethane (TPU) Outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Martino (Frontier 162)
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 43
Toe Material Steel