missing translation for 'onlineSavingsMsg'
Learn More

Maslinic acid, MedChemExpress

Product Code. 16421468
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16421468

Brand: Medchem Express HYN0629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α, which is required for p65 activation.

Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α, which is required for p65 activation.

Specifications

Chemical Name or Material Maslinic acid
CAS 4373-41-5
Color White
Purity Grade Notes Research
Molecular Formula C30H48O4
Quantity 5 mg
Synonym Crategolic acid 2α-Hydroxyoleanolic acid
Solubility Information DMSO : 100 mg/mL (211.55 mM; Need ultrasonic)
SMILES C[C@]([C@@]1([H])[C@]2(C[C@@H](O)[C@@H]3O)C)(CC[C@@]2([H])C3(C)C)[C@@](C4=CC1)(CC[C@]5(C(O)=O)[C@@]4([H])CC(C)(C)CC5)C
Molecular Weight (g/mol) 472.7
Formula Weight 472.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.