missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mass Spectrometry Compatible Crosslinkers

Product Code. 17757463 Shop All Gibco Products
Change view
Click to view available options
Description:
DSPP
TBDSPP
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17757463 TBDSPP 50mg
17747463 DSPP 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17757463 Supplier Thermo Scientific™ Supplier No. A52287
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Improve mass spectrometry detection of crosslinked proteins and interactions with DSPP and TBDSPP heterotrifunctional crosslinkers.

Chemical crosslinking in combination with mass spectrometry is a powerful method to improve protein characterization, and to identify protein-protein interactions. This method has been applied to recombinant and native protein complexes, and more recently, to whole cell lysates or intact unicellular organisms in efforts to identify protein-protein interactions on a global scale. The enrichment feature of DSPP and TBDSPP improves detection of crosslinked proteins by reducing sample complexity through separation of crosslinked from non-crosslinked peptides.

• Tri-functional crosslinker: NHS ester (both ends), phosphonic acid (DSPP), or phosphonate ester (TBDSPP) in the spacer for enrichment
• High-purity crystalline reagents for protein structure and interaction characterization
• Membrane-permeable version (TBDSPP) for intracellular crosslinking
• Enrichable using Fe-NTA IMAC or TiO2 MOAC

DSPP (disuccinimidyl phenyl phosphonic acid, PhoX) and TBDSPP (tert-butyl disuccinimidyl phenyl phosphonate, tBu-PhoX) are phospho-enrichable crosslinkers containing an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of a 7-atom spacer arm with a phosphonic acid group (DSPP) or phosphonate ester (TBDSPP) for enrichment. Both react efficiently with primary amine groups (-NH2) in pH 7–9 buffers to form stable amide bonds. The phosphonic acid or phosphonate ester groups can then be used to enrich crosslinked peptides using immobilized metal affinity chromatography (IMAC) or metal oxide affinity chromatography (MOAC).

Of the two enrichable MS crosslinkers, DSPP is more water soluble than TBDSPP and is better for in vitro crosslinking, simple purified proteins and protein complexes. TBDSPP is better for intracellular crosslinking due to increased membrane permeability. Although both crosslinkers can be enriched using traditional phosphopeptide methods, TBDSPP crosslinked peptides must first be incubated with TFA to remove the tert-butyl protection groups. The TBDSPP phosphonic acid group (after acid cleavage) is not a substrate for phosphatases, which provides opportunity for the removal endogenous phosphopeptides in samples prior to enrichment.

Note: For high resolution analysis of the crosslinked peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164940 or 164568). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES900 or ES904).

TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description TBDSPP
Molecular Weight (g/mol) 552.46
PEGylated No
Purity >90%
Spacer Arm Length 4.3 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability Yes
Product Line MS Reagents
Labeling Method Chemical Labeling
Crosslinker Type Homotrifunctional
Reactive Moiety NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
What is the advantage of using phospho-enrichable crosslinkers?

Phospho-enrichable crosslinkers such as DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) and TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX) are great tools for studying protein-protein Interaction, protein 3D structure, and for mass spectrometry (MS) analysis. They can be used to enrich low-abundance crosslinked peptides and thereby improve MS identification rates.

Is TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX) cell-permeable?

Yes, TBDSPP is cell-permeable. After the crosslinking reaction and before the phospho-enrichment step, TBDSPP needs to be de-protected.

Is DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) cell-permeable?

DSPP is not cell-permeable. We offer TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX), Cat. No. A52287 that is a cell-permeable phopho-enrichable crosslinker.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.