Learn More
Description
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
- Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Label or Dye | Bromophenol Blue |
| Ready to Load | Yes |
| Size Range | 1500 to 10000 bp |
| Gel Compatibility | Agarose Gels |
| Quantity | 2 x 500 μL |
| Product Line | MassRuler |
| Product Type | High DNA Mass Ladder |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.