missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast Cell Protease-11/Prss34 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2857-SP
This item is not returnable.
View return policy
Description
Mast Cell Protease-11/Prss34 Polyclonal specifically detects Mast Cell Protease-11/Prss34 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Mast Cell Protease-11/Prss34 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EOS, Mast Cell Protease11, MCP-11, MCPT11, Prss34 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 260429 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_848459 | |
| Prss34 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-11/Prss34 Met20-Ser318 Accession # NP_848459 | |
| 25 μg | |
| Primary | |
| Detects mouse Mast Cell Protease-11/Prss34 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction