missing translation for 'onlineSavingsMsg'
Learn More

Atago™ MASTER-RI Model Refractometer

Product Code. 11426168
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11426168 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11426168 Supplier Atago™ Supplier No. 2612
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Refractometer
Refractive Index 1.435 to 1.520
Model MASTER-RI
For Use With (Application) Measuriing the refractive index of vegetable oil, petroleum, oil slicks
Weight (Metric) 130 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.