missing translation for 'onlineSavingsMsg'
Learn More

Atago™ MASTER-SUR/Nα Model Clinical Refractometer

Product Code. 11446168
Change view
Click to view available options
Refractive Index:
1.333 to 1.356
1.3330 to 1.3660
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index Unit Size
11446168 1.3330 to 1.3660 Each
11466168 1.333 to 1.356 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446168 Supplier Atago™ Supplier No. 2771
  • ONLINE EXCLUSIVE PRICE
    202.00 €
    183.45 € / Pack of 1
    Save 18.55 €
    9% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Type Clinical Refractometer
Scale Range 1.000 to 1.060 Urine Specific Gravity, 0 to 12 g/100 mL Serum Protein
Refractive Index 1.3330 to 1.3660
Model MASTER-SUR/Nα
Accuracy ±0.001 Urine S.G. scale, ±0.2g/100mL Serum Protein Scale, ±0.0005nD Refractive index scale at 10 to 30°C
Scale Type Urine S.G., Serum protein, Refractive index
For Use With (Application) Designed for clinical purposes: Serum Protein, Specific Gravity of Urine and Refractive Index
IP Rating IP65
Weight (Metric) 110 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.