missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Mastercycler Nexus X2, GB-Plug

Product Code. 15427132 Shop All Eppendorf Products
Change view
Click to view available options
Capacity: 64/32 x 0.2 mL PCR tubes or 24/10 x 0.5 mL PCR tube positions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15427132 Capacity: 64/32 x 0.2 mL PCR tubes or 24/10 x 0.5 mL PCR tube positions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427132 Supplier Eppendorf™ Supplier No. 6337000043
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PCR cycler with the capacity to run two simultaneous and independent protocols.

  • Large block for large assays - small block for small assays
  • Intuitive graphic programming
  • Universal block for strips, 0.1 mL, 0.2 mL, and 0.5 mL PCR tubes
  • Small footprint
  • Optional gradient for PCR optimization
  • E-mail notification
  • flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
TRUSTED_SUSTAINABILITY

Spezifikation

Temperature Range (Metric) 37 to 110°C (Lid), 4 to 99°C (Block)
Content And Storage GB Plug
Dimensions 25 × 41.2 × 33 cm
Capacity 64/32 × 0.2 mL PCR Tubes or 24/10 × 0.5 mL PCR Tube Positions
Electrical Requirements 230 V, 50/60 Hz
For Use With (Application) Standard PCR, Optimizing a PCR
User Interface CAN_in/CAN_out, Ethernet, USB
No. of Programs >700
Heated Lid Yes
Weight 11 kg
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.