Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Mounting Hardware
Product Code. 11731019 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
(1) L/S Easy-Load II pump heads
(2) L/S Easy-Load II pump heads
(3) L/S Easy-Load II pump heads
(4) L/S Easy-Load II pump heads
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11731019 (2) L/S Easy-Load II pump heads Each
11721019 (1) L/S Easy-Load II pump heads Each
11771019 (3) L/S Easy-Load II pump heads Pack of 2
11781019 (4) L/S Easy-Load II pump heads Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11731019 Supplier Masterflex™ Supplier No. 7720002
  • ONLINE EXCLUSIVE PRICE
    86.00 €
    58.15 € / Pack of 1
    Save 27.85 €
    32% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
  • For use with L/S Easy-Load II Pump Heads
  • See individual drive specifications for maximum number of heads to be mounted
  • Mounting hardware is stainless steel
Specifications
For Use With (Application) Mounting L/S Easy-Load II pump heads
For Use With (2) L/S Easy-Load II pump heads
Type Mounting Hardware
Case Material Stainless Steel