Learn More
Masterflex™ Stainless Steel Tubing Weight
Product Code. 11835191
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 11835191
Brand: Masterflex™ 0757004
Only null left
Add to basket
Add to basket
Keep your samples flowing and tubing from curling
Constructed of durable stainless steel
Specifications
For Use With (Application) For groundwater sampling
For Use With For fits Masterflex™ L/S 15 or L/S 24 precision tubing
Material Stainless Steel
Quantity 1 Each
Type Tubing Weight