missing translation for 'onlineSavingsMsg'
Learn More

Corning Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-free, LDEV-free

Product Code. 32226692 Shop All Corning Products
Change view
Click to view available options
Concentration:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
32226692 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32226692 Supplier Corning Supplier No. 6000025301
  • ONLINE EXCLUSIVE PRICE
    426.28 €
    398.08 € / 5mL
    Save 28.20 €
    6% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

  • Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type 2D and 3D
Classification Animal Origin, Reduced Growth Factor
Concentration 5 mL
Form Frozen
Product Type Extracellular Matrix
Serum Level Serum-free
For Use With (Application) Cell Growth and Differentiation, Metabolism and Toxicology Studies, Invasion Assays, in vitro and in vivo Angiogenesis Assays, in vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Manufacturing Quality 9001
Product Line Corning Matrigel
Quantity 5 mL
Shipping Condition -20°C
Assays Angiogenesis, Invasion Assays and drug screening in 3D
Certifications/Compliance Research Use Only
Endotoxin Level >16 EU/mL
Format Frozen
Formulation Dulbecco's Modified Eagle's Medium with 50 μg/mL gentamycin.
Included Antibiotics Gentamycin
Regulatory Status RUO
Source Engelbreth-Holm-Swarm (EHS) Mouse Sarcoma
Species Mouse
Stem Cell Qualified No
Tested For Bacteria, Fungi, Mycoplasma
Color Clear
Content And Storage -20°C
Culture Type 3D Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Adherent Cell Culture, Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.