missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Matrine

Product Code. 16482356
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482356 Supplier MedChemExpress Supplier No. HYN0164
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Matrine (Matridin-15-one) is an alkaloid found in plants from the Sophora genus that can act as a kappa opioid receptor and u-receptor agonist.

Matrine has a variety of pharmacological effects, including anti-cancer, anti-oxidative stress, anti-inflammation and anti-apoptosis effects. Matrine is potential in the research of disease like human non-small cell lung cancer, hepatoma, papillary thyroid cancer and acute kidney injury (AKI).

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 248.36 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 519-02-8
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 50 mg/mL (201.32 mM) ∣H2O : 20 mg/mL (80.53 mM∣Need ultrasonic)
Synonym Matridin-15-one∣ Vegard∣ α-Matrine
Target Opioid Receptor∣Autophagy∣Mitophagy∣Ferroptosis∣Apoptosis
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.