missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes in Barcoded Latch Racks
BIOPHARMAGREENER_CHOICE

Product Code. 15583694
Change view
Click to view available options
Product Type:
Screw Top Tube
Screw Top with White Patch
Color:
Amber
Clear
Closure Color:
Blue
Colorless
Green
Not Included
Purple
Red
White
Yellow
Unit Size:
Pack of 480
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Closure Color Unit Size
15583694 Screw Top Tube Clear Colorless Pack of 480
15553704 Screw Top Tube Clear White Pack of 480
15513704 Screw Top Tube Clear Blue Pack of 480
15573704 Screw Top Tube Clear Not Included Pack of 480
15533704 Screw Top Tube Clear Purple Pack of 480
15523704 Screw Top Tube Clear Green Pack of 480
15503704 Screw Top Tube Amber Red Pack of 480
15543704 Screw Top Tube Clear Red Pack of 480
15543714 Screw Top Tube Amber Not Included Pack of 480
15563704 Screw Top Tube Clear Yellow Pack of 480
15593694 Screw Top with White Patch Clear Colorless Pack of 480
15583704 Screw Top with White Patch Clear Not Included Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15583694 Supplier Thermo Scientific™ Supplier No. 3741BR
  • ONLINE EXCLUSIVE PRICE
    593.00 €
    554.02 € / Pack of 480
    Save 38.98 €
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Track your samples easily at low temperatures, including vapor phase Liquid Nitrogen with 1.0mL ScrewTop Tubes in barcoded latch racks.

Securely store samples at low temperatures, including vapor phase Liquid Nitrogen with these 1.0mL ScrewTop Tubes in barcoded latch racks. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. These tubes are packed in our Barcoded Latch Racks that feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment. The 2D screwtop tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well.

Dependable Sample Protection

  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy ABS to protect tubes in case of drops
  • ScrewTop tubes are compatible with storage down to -196°C

Secure Tracking

  • Permanent, unique 2D tube barcode is laser-etched on bottom of each tube
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking
  • 2D barcode on tubes allow you to scan and decode tubes all at once without removing them from the rack, and allows data to be associated with each tube
  • Tube and rack are compatible with Thermo Scientific™ VisionMate™ readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at once
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Closure Color Colorless
Autoclavable Not Autoclavable
Capacity (Metric) 1.0 mL
Bottom Shape Conical
Material Polypropylene (Medical Grade)
Packaging 5 Latch Racks of 96 Tubes
Barcode Tube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction; Rack barcode: 2D Datamatrix; Linear EAN Code 128; Human Readable
Product Line Matrix
Sterility Sterile
Closure Included Yes
Closure Material Virgin Class VI Medical Grade Polypropylene with Virgin Class VI Medical Grade Silicone O-ring
Capacity (English) 0.0338 fl oz.
Fit for Purpose Sterile and PCR Ready
Product Type Screw Top Tube
Green Features ACT Ecolabel (Gold)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.