Learn More
Thermo Scientific™ Matrix™ Blank and Alphanumeric Storage Tubes
BIOPHARMA
Product Code. 10785204
Change view
Click to view available options
Printed:
Alphanumeric
Blank
Packaging:
10 Latch Racks of 96 Tubes
10 Refill Carriers of 96 Tubes
10 Snap Racks of 96 Tubes
Bulk, 1000 Tubes
Sterility:
Non-sterile
Sterile
Fit for Purpose:
PCR Ready
Sterile and PCR Ready
Unit Size:
Pack of 1000
Pack of 960
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Printed Packaging Sterility Fit for Purpose Unit Size
10785204 Blank 10 Refill Carriers of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
11587399 Alphanumeric 10 Refill Carriers of 96 Tubes Non-sterile PCR Ready Pack of 960
10630784 Blank Bulk, 1000 Tubes Non-sterile PCR Ready Pack of 1000
10111923 Alphanumeric 10 Snap Racks of 96 Tubes Non-sterile PCR Ready Pack of 960
10583055 Blank 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
10620224 Blank Bulk, 1000 Tubes Non-sterile PCR Ready Pack of 1000
10622114 Alphanumeric 10 Latch Racks of 96 Tubes Non-sterile Sterile and PCR Ready Pack of 960
11838433 Blank 10 Snap Racks of 96 Tubes Non-sterile Sterile and PCR Ready Pack of 960
10775204 Blank 10 Latch Racks of 96 Tubes Non-sterile PCR Ready Pack of 960
10661544 Blank 10 Refill Carriers of 96 Tubes Non-sterile PCR Ready Pack of 960
10496163 Alphanumeric 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
10651544 Blank 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
10131493 Alphanumeric 10 Latch Racks of 96 Tubes Non-sterile PCR Ready Pack of 960
10775974 Blank 10 Latch Racks of 96 Tubes Non-sterile PCR Ready Pack of 960
10581325 Alphanumeric 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
11820462 Alphanumeric 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10785204 Supplier Thermo Scientific™ Supplier No. 4250
  • ONLINE EXCLUSIVE PRICE
    192.00 €
    161.10 € / Pack of 960
    Save 30.90 €
    16% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Store valuable samples where 2D tracking is not required with high-density blank and alphanumeric storage tubes.
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or DeepWell™ block sample storage.
Features:
Highest Quality
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Superior Storage Rack Design
  • Unlike traditional storage tubes or blocks, Thermo Scientific Matrix blank and alphanumeric storage tubes are available in our specially designed, stackable, microplate-footprint Latch Racks of 96 tubes for individual sample, access and removal
  • Enables multichannel pipette access and eliminates contamination risk with a lid that never touches the bench top when open
  • Compatible with automated liquid handling, as the lid can be lifted robotically for high-throughput applications
Clear Designations
  • Clearly marked with high contrast printing on the base of each tube, designating their appropriate well location within a rack, for easy identification
  • Enhance ease of visual identification with colored SepraSeal capping options or labelling on the side of the tube using the ID Scribe instrument
Capping Options
  • Several sealing options available, including the Thermo Scientific Matrix line of CapMats and CapStrips for sealing multiple tubes and the Thermo Scientific Matrix SepraSeal and DuraSeal capping systems for individual tube sealing, access and piercing
  • Applied with the Thermo Scientific SuperSealer, sealing is secure and consistent every time
Compatible with:
Capping systems, SepraSeals, Duraseals, SuperSealer, ID Scribe
Recommended for:
Individual sample retrieval; Shipping samples; Storage of DNA, compounds or any other samples
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1.4 mL
Material Polypropylene
Packaging 10 Refill Carriers of 96 Tubes
Product Line Matrix
Sterility Sterile
Capacity (English) 0.0473 fl oz.
Dnase Free Yes
Printed Blank
Fit for Purpose Sterile and PCR Ready
Product Type Storage Tube