missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes
BIOPHARMAGREENER_CHOICE

Product Code. 12831991
Change view
Click to view available options
Closure Color:
Blue
Colorless
Green
Not Included
Purple
Red
White
Yellow
Color:
Amber
Clear
Packaging:
5 Latch Racks of 96 Tubes
Bulk, 480 Tubes
Unit Size:
Pack of 480
Pack of 4800
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Color Packaging Unit Size
12831991 White Clear 5 Latch Racks of 96 Tubes Pack of 480
10725784 Colorless Clear Bulk, 480 Tubes Pack of 480
11661990 Red Clear Bulk, 480 Tubes Pack of 480
11652000 Yellow Clear Bulk, 480 Tubes Pack of 480
11631960 Blue Clear Bulk, 480 Tubes Pack of 480
12811981 Green Clear Bulk, 480 Tubes Pack of 480
11622010 White Clear Bulk, 480 Tubes Pack of 480
11681980 Purple Clear Bulk, 480 Tubes Pack of 480
10089692 Colorless Clear 5 Latch Racks of 96 Tubes Pack of 480
12821991 Red Clear 5 Latch Racks of 96 Tubes Pack of 480
12841991 Yellow Clear 5 Latch Racks of 96 Tubes Pack of 480
12891981 Blue Clear 5 Latch Racks of 96 Tubes Pack of 480
12801991 Green Clear 5 Latch Racks of 96 Tubes Pack of 480
12811991 Purple Clear 5 Latch Racks of 96 Tubes Pack of 480
12336263 Not Included Amber 5 Latch Racks of 96 Tubes Pack of 4800
12326263 Red Amber 5 Latch Racks of 96 Tubes Pack of 480
13426888 Colorless Clear 5 Latch Racks of 96 Tubes Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 12831991 Supplier Thermo Scientific™ Supplier No. 3741WHI
  • ONLINE EXCLUSIVE PRICE
    563.00 €
    481.50 € / Pack of 480
    Save 81.50 €
    14% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Store samples securely at low temperatures, including vapor phase liquid nitrogen, with these 1.0mL screw-top tubes that include color-coded caps to simplify storage.

Securely store samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.

Features:

Dependable Sample Protection

  • Tubes are constructed of virgin class VI medical grade polypropylene
  • Screw-top caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • Screw-top tubes are compatible with storage down to -196°C

Secure Tracking

  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping/decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time

Stringent Quality Control

  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All tubes are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Closure Color White
Autoclavable Not Autoclavable
Capacity (Metric) 1.0 mL
Bottom Shape Conical
Material Polypropylene (Medical Grade)
Packaging 5 Latch Racks of 96 Tubes
Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
Product Line Matrix
Sterility Sterile
Closure Included Yes
Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket
Capacity (English) 0.0338 fl oz.
Dnase Free Yes
Fit for Purpose Sterile and PCR Ready
Product Type Screw Top Tube
Green Features ACT Ecolabel (Gold)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.