missing translation for 'onlineSavingsMsg'
Learn More

3M™ Matting Adhesive

Product Code. 17138750
Change view
Click to view available options
Color:
Orange
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17138750 Orange Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17138750 Supplier 3M™ Supplier No. 7100133178
  • ONLINE EXCLUSIVE PRICE
    175.00 €
    87.12 € / Pack of 12
    Save 87.88 €
    50% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Urethane-based matting adhesive for use with 3M™ Seaming Tape and/or 3M™ Matting Edging

  • For seaming & attaching edging to vinyl or vinyl-backed matting
  • Use with 3M™ Matting Seaming Tape to join vinyl mats together to produce a custom-sized mat
  • Also for use with 3M™ Matting Edging to provide tapered edges to vinyl or vinyl-backed matting
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Seaming and attaching edging to Vinyl or Vinyl-backed matting
Quantity 88.7 mL
Color Orange
Type Matting Adhesive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.