Learn More
Honeywell™ Max Single-Use Earplug LS-400 Refill
Product Code. 12656123
Change view
Click to view available options
Orange, 200 Pairs/Pk
Product Code.
12656123 Orange, 200 Pairs/Pk
1 options
Product Code. 12656123 Supplier Honeywell™ Supplier No. 1013046
  • ONLINE EXCLUSIVE PRICE
    79.75 €
    44.18 € / Pack of 200
    Save 35.57 €
    44% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Soft foam earplugs provide superior long-wearing comfort and solid performance.
  • Soft foam for comfort and performance
  • Bell-shaped design for maximum in-ear comfort without backing out of the ear canal, easier to insert
  • Smooth, soil-resistant surface prevents dirt or other substances from accumulating on skin or entering ear canal
  • Ideal for workers who need to block out loud noise and demand the best protection and comfort available in an earplug
  • Europe's most used polyurethane earplug
  • Superior noise-blocking performance in a single-use earplug
  • Enhanced, contoured shape is easier to insert and stays in for long-wearing comfort
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Certifications/Compliance (Europe) EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: 1219 EC Attestation: 04.03.66
Pairs per Package 200 Pairs/Pk.
Noise Reduction Rating (NRR) SNR: 37 dB
Plug Shape Bell
Product Type Single-use earplug, dispenser refill
Disposable Single-use
Material Polyurethane