missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima™ H Minus cDNA Synthesis Master Mix

Product Code. 15686019 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Kit only
Kit with dsDNase
No. of Reactions:
200 Reactions
50 Reactions
Unit Size:
200 reactions
50 reactions
Product Code. Includes No. of Reactions unitSize
15686019 Kit only 200 Reactions 200 reactions
15676019 Kit only 50 Reactions 50 reactions
15696019 Kit with dsDNase 50 Reactions 50 reactions
15606029 Kit with dsDNase 200 Reactions 200 reactions
4 options
This item is not returnable. View return policy

Product Code. 15686019

Brand: Thermo Scientific™ M1662

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Thermo Scientific Maxima H Minus cDNA Synthesis Master Mix, available with or without dsDNase, provides all cDNA synthesis reaction components in a convenient one-tube master mix.

Thermo Scientific Maxima H Minus cDNA Synthesis Master Mix, available with or without dsDNase, provides all cDNA synthesis reaction components in a convenient one-tube master mix. It is optimized for highly efficient cDNA synthesis for two-step quantitative RT-PCR (RT-qPCR) applications and reproducible cDNA synthesis at elevated temperatures (50–65°C) within 30 minutes.

The master mix contains Maxima H Minus Reverse Transcriptase (RT) and RiboLock RNase Inhibitor. Maxima H Minus RT is an advanced enzyme derived from M-MuLV RT by in vitro evolution. The enzyme features high thermostability, robustness, processivity, increased cDNA synthesis rate, and lack of RNase H activity. The recombinant RiboLock RNase Inhibitor effectively protects RNA template from degradation by RNases A, B, and C at temperatures up to 55°C. The master mix also contains reaction buffer, dNTPs, oligo (dT)18, and random hexamer primers. A separate tube of No-RT control mix is also provided for convienient RT negative control.

Both kits are capable of reproducible cDNA synthesis at elevated temperatures (50°C–65°C). The synthesis reaction is typically complete in 15–30 minutes. The included double-strand specific DNase in the Maxima H Minus cDNA Synthesis Master Mix with dsDNase allows removal of genomic DNA from RNA samples in two minutes without affecting the quality or quantity of RNA. This dsDNase is also available separately (Cat. No. EN0771).

Features of Maxima H Minus cDNA Synthesis Master Mix include:
• One-tube master mix helps reduce pipetting steps and enhanced consistency in RT-qPCR results
• Increased RT efficiency across a wide range of input RNA amounts and gene targets
• High thermostability to allow RT reactions at 50–65°C temperature range
• Integrated step of genomic DNA removal from RNA samples (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)

Additional information about reaction components
• The No RT control mix contains all the components in the Maxima H Minus cDNA Synthesis Master Mix except RT. The presence of a PCR product in the No RT control reaction indicates that the reaction is contaminated with DNA. To further enhance genomic DNA elimination efficiency, template RNA incubation with dsDNase (Cat. No. EN0771) is strongly recommended.
• Nuclease-free water is provided for reaction setup and dilution of sample DNA. The absence of exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
• To further enhance genomic DNA elimination efficiency, template RNA incubation with the included dsDNase is strongly recommended. (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)
• The dsDNase is used for rapid and safe removal of contaminating genomic DNA from RNA samples. dsDNase is easily inactivated by moderate heat treatment (55°C). It allows for a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. (Maxima H Minus cDNA Synthesis Master Mix with dsDNase only)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage

• Maxima H Minus cDNA Synthesis Master Mix (1 x 800 μL)
• Maxima H Minus cDNA Synthesis Master Mix 'No RT' Control (1 x 200 μL)
• Nuclease-free water (1 x 1.25 mL)

Store at –5 to –30°C.

Format Master Mix
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity Each
Reverse Transcriptase Maxima H Minus
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 200 Reactions
Reaction Format Premixed Components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Includes Kit only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.