missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima H Minus Double-Stranded cDNA Synthesis Kit

Product Code. 13293159 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Reactions
200 Reactions
50 Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13293159 10 Reactions 10 reactions
16652247 50 Reactions 50 reactions
17164891 200 Reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13293159 Supplier Thermo Scientific™ Supplier No. K2561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

First Strand Enzyme Mix, 4X First Strand Reaction Mix, Second Strand Enzyme Mix, 5X Second Strand Reaction Mix, 0.5M EDTA ( pH 8.0), RNase I (10 u/μL), Control RNA (0.5μg/μL), Oligo(dT)18 Primer, 100μM, 0.5μg/μL, Random Hexamer Primer, 100μM, 0.2μg/μL,Water, nuclease-free, Detailed Protocol

The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.

Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA. First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains pre-mixed components to reduce the number of pipetting steps necessary to complete the procedure.

Features of the Maxima H Minus Double-Stranded cDNA Synthesis Kit include:

  • Efficient synthesis of full-length double-stranded cDNA
  • Fast—procedure completed in less than two hours
  • Convenient—pre-mixed components
  • Complete—includes all primers, controls and residual RNA removal reagents

Applications

  • Full-length double-stranded blunt-end cDNA synthesis for cloning
  • Double-stranded cDNA library construction

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY

Specifications

Format 10 reactions
Optimal Reaction Temperature 50°-55° C
Product Line Maxima
Product Type cDNA Synthesis Kit
Quantity 10 Reactions
Reverse Transcriptase Maxima H Minus
For Use With (Application) Cloning
Final Product Type Double Stranded cDNA
No. of Reactions 10 Reactions
Reaction Format Separate Components
Sample Type RNA
Show More Show Less
compliance-icons
Product Identifier
  • Maxima H Minus Double-Stranded cDNA Synthesis Kit
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: EDTA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.