missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima H Minus Double-Stranded cDNA Synthesis Kit

Product Code. 16652247
Change view
Click to view available options
Quantity:
10 Reactions
200 Reactions
50 Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16652247 50 Reactions 50 reactions
17164891 200 Reactions 200 reactions
13293159 10 Reactions 10 reactions
15707536 10 Reactions 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16652247 Supplier Thermo Scientific™ Supplier No. K2562
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.

Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA. First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains pre-mixed components to reduce the number of pipetting steps necessary to complete the procedure.

Features of the Maxima H Minus Double-Stranded cDNA Synthesis Kit include:

  • Efficient synthesis of full-length double-stranded cDNA
  • Fast—procedure completed in less than two hours
  • Convenient—pre-mixed components
  • Complete—includes all primers, controls and residual RNA removal reagents

Applications

  • Full-length double-stranded blunt-end cDNA synthesis for cloning
  • Double-stranded cDNA library construction

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -25°C to -15°C
Format 50 reactions
Optimal Reaction Temperature 50°C to 55°C
Product Line Maxima
Product Type cDNA Synthesis Kit
Quantity 50 Reactions
Reverse Transcriptase Maxima H Minus
For Use With (Application) Cloning
Final Product Type Double Stranded cDNA
No. of Reactions 50 Reactions
Reaction Format Separate Components
Sample Type RNA
Show More Show Less
What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.