Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
Brand:  Thermo Scientific™ K1681
Additional Details : Weight : 1.02060kg
 View more versions of this product
Product Code. 15787526
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
  • High yields of full-length first strand cDNA -with RNA templates up to 20 kb
  • Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Specifications
Specifications
RT-PCR
20 x 20μL Reactions