Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
134.00 € - 482.00 €
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15777526
|
Thermo Scientific™
K1652 |
100 reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15787526
|
Thermo Scientific™
K1681 |
20 x 20μL Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
- High yields of full-length first strand cDNA -with RNA templates up to 20 kb
- Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Specifications
RT-PCR |