missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kit

Product Code. 15235146 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Kit only
Kit with dsDNase
No. of Reactions:
100 Reactions
20 Reactions
Unit Size:
100 reactions
20 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes No. of Reactions unitSize
15235146 Kit with dsDNase 100 Reactions 100 reactions
10338179 Kit only 20 Reactions 20 reactions
10430470 Kit only 100 Reactions 100 reactions
15264071 Kit with dsDNase 20 Reactions 20 reactions
15797526 Kit with dsDNase 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 15235146 Supplier Thermo Scientific™ Supplier No. K1682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA.

Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA. The kit uses the Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT. The enzyme features the highest thermostability among the derivatives of MMLV RT and lacks RNase H activity.

This kit allows synthesis of long cDNAs up to 20 kb at elevated temperatures (up to 65°C), superseding other systems' abilities to produce full-length cDNA. Due to increased synthesis rates the reaction can be completed in 30 minutes.

For reverse transcription the kit uses Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT.

The Maxima H Minus First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand-specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific dsDNase activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved, and dsDNase treated RNA can be directly added to reverse transcription.

Features of the Maxima H Minus First Strand cDNA Synthesis Kit include:

  • Increased reaction temperatures—the first strand of cDNA can be synthesized within the 42–65°C temperature range
  • High yields of full-length first strand cDNA—with RNA templates up to 20 kb
  • Flexible priming—oligo(dT)18, random hexamer or gene-specific primers
  • Integrated genomic DNA removal step with dsDNase kit

Applications

  • First Strand cDNA synthesis for RT-PCR
  • Construction of cDNA libraries
  • Generation of probes for hybridization
  • Antisense RNA synthesis

Contents

  • Maxima H Minus Enzyme Mix
  • Oligo(dT)18 and Random hexamer primers
  • 5X RT Buffer
  • dNTP Mix
  • Nuclease-free water

First Strand Synthesis Kit with dsDNase also contains

  • dsDNase and 10X dsDNase Buffer

Additional information about reaction components

  • Maxima H Minus Enzyme Mix contains Maxima H Minus Reverse Transcriptase and RiboLock RNase Inhibitor. RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
  • Oligo(dT)18 and random hexamer primers are supplied with the kit. Random hexamer primers bind non-specifically and are used to synthesize cDNA from all RNAs in a total RNA population. The oligo(dT)18 primer selectively anneals to the 3'-end of poly(A) RNA, synthesizing cDNA only from poly(A) tailed mRNA. Gene-specific primers may also be used with the kit to prime synthesis from a specified sequence.
  • 10 mM dNTP Mix is a premixed aqueous solution of dATP, dTTP, dCTP, and dGTP.
  • Nuclease-free water is provided for reaction set-up and dilution of sample DNA. The absence of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• Maxima H Minus Enzyme Mix
• dsDNase
• 10X dsDNase Buffer
• Oligo(dT)18 and Random hexamer primers
• 5X RT Buffer
• dNTP Mix
• Nuclease-free water

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C to 55°C
Quantity Each
Reverse Transcriptase Maxima H Minus
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 100 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Includes Kit with dsDNase
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.