missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima Reverse Transcriptase (200 U/μL)

Product Code. 10869200 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10,000 Units
2,000 Units
4 x 10,000 Units
Unit Size:
10000 units
2000 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10869200 10,000 Units 10000 units
10899010 2,000 Units 2000 units
10420100 4 x 10,000 Units 2000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10869200 Supplier Thermo Scientific™ Supplier No. EP0742
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity as well as RNase H activity.

Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity as well as RNase H activity. The engineered enzyme features dramatically improved thermostability and robustness, and increased synthesis rates compared to wild type M-MuLV RT.

Maxima Reverse Transcriptase is capable of reproducible cDNA synthesis from a wide range of input total RNA amounts (from 1 pg to 5 μg) at elevated temperatures (50–65°C), which makes this enzyme an ideal tool for two step RT-qPCR (see supporting data).

Due to its high thermostability, Maxima enzyme maintains full activity during the entire reverse transcription reaction, generates the highest yields of cDNA, and is able to synthesize even very long RNA transcripts up to 20 kb. The reaction temperature can be increased up to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. Due to its increased synthesis rate, the reverse transcriptase reaction can be completed in 15 to 30 minutes.

Features of Maxima Reverse Transcriptase include:

  • High yields of full-length cDNA up to 20 kb
  • Active up to 65°C
  • Thermostable—90% active after incubation at 50°C for 60 minutes
  • High sensitivity allows reproducible cDNA synthesis from a wide range of total RNA quantities (1 pg to 5 μg)
  • Efficient formula for complete cDNA synthesis in 15 to 30 minutes
  • Incorporates modified nucleotides

Applications

  • Full-length cDNA synthesis for long transcripts and ORFs
  • Reverse transcription at elevated temperatures to reduce effects of secondary structure
  • cDNA cloning and construct generation
  • RNA-seq & single cell RNA-seq library prep
  • Isoform and transcript characterization
  • Template-switching-based full-length RNA-seq

Also available

  • Maxima Reverse Transcriptase (Cat. No. EP0741)
  • Maxima First Strand cDNA Synthesis Kit for RT-qPCR (Cat. No. K1641)
  • Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase (Cat. No. K1671)
TRUSTED_SUSTAINABILITY

Specifications

Concentration 200 U/μL
Content And Storage

• Maxima Reverse Transcriptase, 1 x 10,000 units (200 U/μL)
• 5X RT Buffer

Store at –20°C.

Format Stand-alone Enzyme
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C to 55°C
Quantity 10,000 Units
Reverse Transcriptase Maxima
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
Final Product Type First-Strand cDNA
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.