Learn More
Maximum Recovery Diluent Ready Bag
Description
Maintain viability and maximize the recovery of microorganisms with protective and isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent Ready Bag. Maximum Recovery Diluent Ready Bags are designed for use in food testing, by laboratories with large numbers of samples, requiring large volumes of broth or diluent.
Shorten your workflow with easy-to-use Maximum Recovery Diluent Ready Bag. Ready to go when you are, the bags are highly adaptable to laboratory workflows and automated equipment, such as dilutors, with no further supplementation required.
- Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.
- Easy to use: easy to dispense with minimum training
- Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste
For added convenience, it is recommended that Ready Bag Enrichment Media be dispensed using the Thermo Scientific Automated Diluter (DB4150A).
Specifications
Specifications
| Color | Colorless, clear |
| Content And Storage | 2 to 25°C |
| Description | Maximum Recovery Diluent Ready Bag |
| Format | 3L bag with connection ports in a box |
| Product Type | Dehydrated Culture Media |
| Fill Weight | 3020mL ± 20mL |
| Quantity | 4/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.