missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Borosilicate Glass Class A Col Lisse

Calibrated to Class A tolerances. MBL™ Borosilicate Glass Class A Col Lisse is iIndividually laser etched with serial number, batch code and date of manufacture

Brand:  MBL FRD212

102.60 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Code : A0

Additional Details : Weight : 0.57000kg

 View more versions of this product

Product Code. 12633272

  • 108.00 € / Pack of 5
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Complies with ISO 1042 and DIN 12664
  • Finely calibrated to Class A tolerances
  • Each flask is laser etched with an individual serial number, batch code and date of manufacture
  • Manufactured from chemically resistant borosilicate glass
  • Conformity 'H' certified
  • Inscriptions in blue enamel
  • Supplied with polyethylene stopper
  • Batch certificate available to download online
  • Complies with ISO 1042
  • With plain neck to accept rubber stopper
  • Manufactured from borosilicate glass
  • Finely calibrated to Class A tolerances
  • Conformity 'H' certified
  • Individually laser etched with serial number, batch code and date of manufacture
  • Batch certificate available to download online
  • Blue enamel inscriptions
Specifications

Specifications

Class A, ISO 1042, Conformity 'H' certified
200 mL
210 mm
±0.15 mL
A
Rubber
Borosilicate Glass
Plain
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers