Learn More
MBL™ Borosilicate Glass Class A Col Lisse
Calibrated to Class A tolerances. MBL™ Borosilicate Glass Class A Col Lisse is iIndividually laser etched with serial number, batch code and date of manufacture
44.65 € - 205.00 €
Specifications
Certifications/Compliance | Class A, ISO 1042, Conformity 'H' certified |
---|---|
Closure Material | Rubber |
Material | Borosilicate Glass |
Neck Style | Plain |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Tolerance | Price | Quantity & Availability | |||
12524927
|
MBL
FRD200 |
5 mL | 70 mm | ±0.25 mL |
49.55 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12683262
|
MBL
FRD202 |
10 mL | 90 mm | ±0.025 mL |
54.00 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12693262
|
MBL
FRD204 |
20 mL | 110 mm | ±0.04 mL |
59.05 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12200353
|
MBL
FRD206 |
25 mL | 110 mm | ±0.04 mL |
74.70 €
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
12613272
|
MBL
FRD208 |
50 mL | 140 mm | ±0.6 mL |
63.20 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12200363
|
MBL
FRD210 |
100 mL | 170 mm | ±0.1 mL |
79.90 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12633272
|
MBL
FRD212 |
200 mL | 210 mm | ±0.15 mL |
108.00 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12200373
|
MBL
FRD214 |
250 mL | 220 mm | ±0.15 mL |
44.65 €
Pack of 2 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12200383
|
MBL
FRD216 |
500 mL | 260 mm | ±0.25 mL |
53.70 €
Pack of 2 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
11930316
|
MBL
FRD218 |
1000 mL | 300 mm | ±0.4 mL |
78.55 €
Pack of 2 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
11940316
|
MBL
FRD220 |
2000 mL | 370 mm | ±0.6 mL |
52.75 €
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
12124432
|
MBL
FRD222 |
5 L | 475 mm | ±1.2 mL |
205.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
Description
- Complies with ISO 1042
- With plain neck to accept rubber stopper
- Manufactured from borosilicate glass
- Finely calibrated to Class A tolerances
- Conformity 'H' certified
- Individually laser etched with serial number, batch code and date of manufacture
- Batch certificate available to download online
- Blue enamel inscriptions
Specifications
Class A, ISO 1042, Conformity 'H' certified | |
Borosilicate Glass | |
Yes |
Rubber | |
Plain | |
A |