missing translation for 'onlineSavingsMsg'
Learn More

MBL™ One Mark Class B Glass Pipets

Product Code. 12693032
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Tolerance:
±0.015 mL
±0.02 mL
±0.03 mL
±0.04 mL
±0.06 mL
±0.1 mL
±0.15 mL
Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Pack of 2
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tolerance Color unitSize
12693032 50 mL ±0.1 mL Red Pack of 5
12510026 100 mL ±0.15 mL Yellow Pack of 2
12580036 10 mL ±0.04 mL Red Pack of 5
12653032 1 mL ±0.015 mL Blue Pack of 5
12673032 20 mL ±0.06 mL Yellow Pack of 5
12633072 25 mL ±0.06 mL Blue Pack of 5
12560036 2 mL ±0.02 mL Orange Pack of 5
12570036 5 mL ±0.03 mL White Pack of 5
12663032 15 mL ±0.06 mL Green Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12693032 Supplier MBL Supplier No. PRA714B
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from soda lime glass with tooled jets to reduce breakage and complies with ISO 648 and DIN 12690. MBL™ One Mark Class B Glass Pipets incude markings in permanent amber stain which are diffused into the surface of the glass for increased chemical resistance. Calibrated for delivery.

  • Complies with ISO 648 and DIN 12690
  • Calibrated for delivery
  • Manufactured from soda lime glass with tooled jets to reduce breakage
  • Markings in permanent amber stain which are diffused into the surface of the glass for increased chemical resistance
TRUSTED_SUSTAINABILITY

Specifications

Calibration To Deliver
Certifications/Compliance ISO 648, DIN 12690
Class B
Material Soda Lime Glass
Graduated Yes
Graduation Color Amber
Quantity 5 Pack
Tolerance ±0.1 mL
Color Red
Volume (Metric) 50 mL
Type Bulb Pipet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.