missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MBPTrap™ HP Prepacked Columns

Product Code. 10097924 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
1mL
Each
Pack of 5
Product Code. Quantity Flow Rate unitSize
10097924 1 x 5mL <20 mL/min. Each
10075174 5 x 1mL <4 mL/min. Pack of 5
11500664 5 x 5mL <20 mL/min. Pack of 5
15127956 1 x 1mL <4 mL/min. 1mL
4 options
This item is not returnable. View return policy

Product Code. 10097924

Brand: Cytiva 28918779

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Yield highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes. Cytiva Life Sciences™ MBPTrap™ HP Prepacked Columns are prepacked with Dextrin Sepharose™ High Performance.

  • Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
  • Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
  • Maximum pressure: 3 bar
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type Dextrin Sepharose HP prepacked column
Bed Volume 1mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type Dextrin
Max. Pressure 3 bar
pH Range >7
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Purification of MBP tagged Proteins
Show More Show Less
compliance-icons
Product Identifier
  • MBPTrap HP, 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.